Search
Search
| Gene Symbol: MBOAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404426.1 | XP_022260134.1 | 1107 | 138 | ||
| XM_022404427.1 | XP_022260135.1 | 1099 | 138 | |||
| XM_022404428.1 | XP_022260136.1 | 762 | 138 | |||
| XM_022404429.1 | XP_022260137.1 | 1225 | 138 | |||
Set Membership: |
|