Search
Search
| Gene Symbol: FBXL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630981.2 | XP_005631038.1 | 1327 | 98 | ||
| XM_005630982.2 | XP_005631039.1 | 1317 | 98 | |||
| XM_005630983.2 | XP_005631040.1 | 1327 | 98 | |||
| XM_005630984.2 | XP_005631041.1 | 1327 | 98 | |||
| XM_005630985.2 | XP_005631042.1 | 1275 | 98 | |||
| XM_005630986.2 | XP_005631043.1 | 1327 | 98 | |||
| XM_022405233.1 | XP_022260941.1 | 1327 | 98 | |||
| XM_022405234.1 | XP_022260942.1 | 1108 | 98 | |||
| XM_022405235.1 | XP_022260943.1 | 1327 | 98 | |||
| XM_022405236.1 | XP_022260944.1 | 994 | 98 | |||
Set Membership: |
|