Search
Search
| Gene Symbol: ENDOV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624018.2 | XP_005624075.1 | 285 | 103 | ||
| XM_022422637.1 | XP_022278345.1 | 285 | 103 | |||
| XM_022422638.1 | XP_022278346.1 | 285 | 103 | |||
| XM_022422639.1 | XP_022278347.1 | 285 | 103 | |||
| XM_022422640.1 | XP_022278348.1 | 285 | 103 | |||
Set Membership: |
|