Search
Search
| Gene Symbol: TMEM65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627911.3 | XP_005627968.1 | 724 | 165 | ||
| XM_005627912.3 | XP_005627969.1 | 925 | 165 | |||
| XM_022426689.1 | XP_022282397.1 | 730 | 150 | |||
| XM_846638.5 | XP_851731.1 | 713 | 165 | |||
Set Membership: |
|