Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Cf02730159_m1
          See other RAPGEF2 GE Assays ›
          Gene Symbol:
          RAPGEF2
          Gene Name:
          Rap guanine nucleotide exchange factor 2
          Gene Aliases:
          CNrasGEF, NRAPGEP, PDZ-GEF1, PDZGEF1, RA-GEF-1
          Chromosome Location:
          Chr.15: 56133094 - 56353609 on Build CanFam3.1
          Species:
          Dog
          Species Specific ID (Flybase ID):
          -
          Assay ID Cf02730159_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RAPGEF2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003639497.4 XP_003639545.1 2 - 3 195 115
          XM_005629386.3 XP_005629443.1 2 - 3 282 115
          XM_014119398.2 XP_013974873.1 5 - 6 559 115
          XM_014119399.2 XP_013974874.1 5 - 6 542 115
          XM_022403554.1 XP_022259262.1 8 - 9 699 115
          XM_022403555.1 XP_022259263.1 7 - 8 681 115
          XM_022403556.1 XP_022259264.1 8 - 9 699 115
          XM_022403557.1 XP_022259265.1 8 - 9 699 115
          XM_022403558.1 XP_022259266.1 8 - 9 699 115
          XM_022403559.1 XP_022259267.1 2 - 3 195 115

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          neuron migration
          G-protein coupled receptor signaling pathway
          neuropeptide signaling pathway
          negative regulation of cell proliferation
          positive regulation of neuron projection development
          cAMP-mediated signaling
          ventricular system development
          forebrain neuron development
          neuron projection development
          brain-derived neurotrophic factor receptor signaling pathway
          positive regulation of protein binding
          Rap protein signal transduction
          nerve growth factor signaling pathway
          positive regulation of GTPase activity
          positive regulation of cAMP-mediated signaling
          positive regulation of protein kinase activity
          negative regulation of melanin biosynthetic process
          negative regulation of dendrite morphogenesis
          establishment of endothelial barrier
          positive regulation of ERK1 and ERK2 cascade
          cellular response to cAMP
          cellular response to cGMP
          adenylate cyclase-activating adrenergic receptor signaling pathway
          regulation of cell junction assembly
          cellular response to nerve growth factor stimulus
          positive regulation of cAMP-dependent protein kinase activity
          positive regulation of dendritic cell apoptotic process
          positive regulation of vasculogenesis
          positive regulation of neuron migration
          Ras guanyl-nucleotide exchange factor activity
          GTPase activator activity
          protein binding
          Rap guanyl-nucleotide exchange factor activity
          PDZ domain binding
          cAMP binding
          beta-1 adrenergic receptor binding
          WW domain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline