Search Thermo Fisher Scientific
Gene Symbol: GPX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115119.1 | NP_001108591.1 | 2 | 358 | 81 | |
GenBank mRNA | AY572225.1 | - | 2 | 298 | 81 | |
DN270560.1 | - | 2 | 400 | 81 | ||
DN870414.1 | - | 2 | 274 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |