Search
Search
| Gene Symbol: SLC4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289433.1 | NP_001276362.1 | 19 - 20 | 3331 | 70 | |
| NM_001289434.1 | NP_001276363.1 | 19 - 20 | 3304 | 70 | ||
| NM_001289435.1 | NP_001276364.1 | 15 - 16 | 2729 | 70 | ||
| GenBank mRNA | HQ379706.1 | - | 19 - 20 | 3297 | 70 | |
| HQ379707.1 | - | 19 - 20 | 3270 | 70 | ||
| HQ379708.1 | - | 15 - 16 | 2729 | 70 | ||
Set Membership: |
|