Search
Search
| Gene Symbol: TRIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628350.3 | XP_005628407.1 | 8 | 1386 | 143 | |
| XM_014119171.2 | XP_013974646.1 | 6 | 1317 | 143 | ||
| XM_014119172.2 | XP_013974647.1 | 5 | 1221 | 143 | ||
| XM_022427182.1 | XP_022282890.1 | 6 | 1308 | 143 | ||
| XM_022427183.1 | XP_022282891.1 | 8 | 1415 | 143 | ||
| XM_539323.6 | XP_539323.3 | 7 | 1300 | 143 | ||
Set Membership: |
|