Search
Search
| Gene Symbol: NPHP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135788.1 | NP_001129260.1 | 2598 | 56 | ||
| XM_005620413.2 | XP_005620470.1 | 2769 | 56 | |||
| XM_014113843.2 | XP_013969318.1 | 2875 | 56 | |||
| XM_014113848.2 | XP_013969323.1 | 2773 | 56 | |||
| XM_022418039.1 | XP_022273747.1 | 2947 | 56 | |||
| XM_022418040.1 | XP_022273748.1 | 2943 | 56 | |||
| XM_022418041.1 | XP_022273749.1 | 2947 | 56 | |||
| XM_022418042.1 | XP_022273750.1 | 2948 | 56 | |||
| XM_022418043.1 | XP_022273751.1 | 2944 | 56 | |||
| XM_022418044.1 | XP_022273752.1 | 2948 | 56 | |||
| XM_022418045.1 | XP_022273753.1 | 2836 | 56 | |||
| XM_022418046.1 | XP_022273754.1 | 2764 | 56 | |||
| XM_022418047.1 | XP_022273755.1 | 2949 | 56 | |||
| XM_022418048.1 | XP_022273756.1 | 2949 | 56 | |||
| GenBank mRNA | EU659993.1 | - | 2598 | 56 | ||
Set Membership: |
|