Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197087.1 | NP_001184016.1 | 1 - 2 | 79 | 74 | |
GenBank mRNA | FJ396442.1 | - | 1 - 2 | 67 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |