Search
Search
| Gene Symbol: KLK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197158.1 | NP_001184087.1 | 1 - 2 | 91 | 94 | |
| XM_005615761.3 | XP_005615818.1 | 2 - 3 | 226 | 94 | ||
| GenBank mRNA | GQ303578.1 | - | 1 - 2 | 91 | 94 | |
Set Membership: |
|