Search
Search

PALM2AKAP2| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204899.1 | NP_001191828.1 | 3 - 4 | 258 | 93 | |
| XM_005626411.3 | XP_005626468.2 | 4 - 5 | 326 | 93 | ||
| XM_005626413.3 | XP_005626470.2 | 4 - 5 | 326 | 93 | ||
| XM_022424914.1 | XP_022280622.1 | 4 - 5 | 339 | 93 | ||
| XM_022424915.1 | XP_022280623.1 | 4 - 5 | 335 | 93 | ||
| XM_022424916.1 | XP_022280624.1 | 4 - 5 | 339 | 93 | ||
| XM_022424917.1 | XP_022280625.1 | 4 - 5 | 326 | 93 | ||
Set Membership: |
|