Search
Search
| Gene Symbol: RPLP0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252137.1 | NP_001239066.1 | 2 - 3 | 284 | 90 | |
| XM_005636025.3 | XP_005636082.1 | 3 - 4 | 439 | 90 | ||
| XM_022410154.1 | XP_022265862.1 | 2 - 3 | 464 | 90 | ||
| GenBank mRNA | GR900033.1 | - | 3 - 4 | 306 | 90 | |
Set Membership: |
|