Search
Search
| Gene Symbol: MIR7-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049423.1 | 1 | 41 | 78 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fragile histidine triad diadenosine triphosphatase | FHIT | Anti-sense | Chr. 20 - 29735380 - 31113859 on NCBI Build CanFam3.1 | ||
| Downstream | CFAP20 domain containing | CFAP20DC | C20H3orf67 | 1.9mb | Anti-sense | Chr. 20 - 31702641 - 31946195 on NCBI Build CanFam3.1 |
Set Membership: |
|