Search
Search

MIR8817| Gene Symbol: MIR8817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128758.1 | 1 | 79 | 81 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | marker of proliferation Ki-67 | MKI67 | -654.5kb | Sense | Chr. 28 - 36920679 - 36939336 on NCBI Build CanFam3.1 | |
| Downstream | O-6-methylguanine-DNA methyltransferase | MGMT | 610.7kb | Anti-sense | Chr. 28 - 38204611 - 38427482 on NCBI Build CanFam3.1 |
Set Membership: |
|