Search
Search

Mef2d| Gene Symbol: Mef2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502908.2 | XP_003502956.1 | 6 - 7 | 909 | 103 | |
| XM_007643523.1 | XP_007641713.1 | 5 - 6 | 805 | 103 | ||
| XM_007643524.1 | XP_007641714.1 | 6 - 7 | 909 | 103 | ||
| XM_007643525.1 | XP_007641715.1 | 6 - 7 | 906 | 103 | ||
| XM_007643526.1 | XP_007641716.1 | 5 - 6 | 771 | 103 | ||
| GenBank mRNA | JI869167.1 | - | 839 | 103 | ||
| JP062042.1 | - | 838 | 103 | |||
Set Membership: |
|