Search
Search

Asic4| Gene Symbol: Asic4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510597.2 | XP_003510645.1 | 8 - 9 | 1398 | 63 | |
| XM_007651219.1 | XP_007649409.1 | 7 - 8 | 1686 | 63 | ||
Set Membership: |
|