Search
Search

Rc3h1| Gene Symbol: Rc3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497667.2 | XP_003497715.1 | 1 - 2 | 421 | 63 | |
| XM_007615590.1 | XP_007613780.1 | 1 - 2 | 421 | 63 | ||
| XM_007615600.1 | XP_007613790.1 | 1 - 2 | 421 | 63 | ||
Set Membership: |
|