Search
Search

Kpna3| Gene Symbol: Kpna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500498.2 | XP_003500546.1 | 11 - 12 | 973 | 75 | |
| GenBank mRNA | JI865750.1 | - | 11 - 12 | 988 | 75 | |
| JP058082.1 | - | 11 - 12 | 957 | 75 | ||
Set Membership: |
|