Search
Search

Nelfb| Gene Symbol: Nelfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513705.2 | XP_003513753.1 | 3 - 4 | 568 | 63 | |
| GenBank mRNA | JI891067.1 | - | 3 - 4 | 295 | 63 | |
| JP052608.1 | - | 3 - 4 | 295 | 63 | ||
Set Membership: |
|