Search
Search

Tbl1xr1| Gene Symbol: Tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510188.2 | XP_003510236.1 | 10 - 11 | 1057 | 64 | |
| XM_007650816.1 | XP_007649006.1 | 10 - 11 | 1110 | 64 | ||
| GenBank mRNA | JI867614.1 | - | 6 - 7 | 516 | 64 | |
| JP064308.1 | - | 6 - 7 | 516 | 64 | ||
Set Membership: |
|