Search
Search

Ust| Gene Symbol: Ust | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498947.2 | XP_003498995.1 | 7 - 8 | 974 | 60 | |
| GenBank mRNA | JI896555.1 | - | 4 - 5 | 449 | 60 | |
| JP038115.1 | - | 4 - 5 | 449 | 60 | ||
Set Membership: |
|