Search
Search

Ctnnd1| Gene Symbol: Ctnnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646633.1 | XP_007644823.1 | 12 - 13 | 2347 | 91 | |
| XM_007646634.1 | XP_007644824.1 | 12 - 13 | 2347 | 91 | ||
| GenBank mRNA | JI892570.1 | - | 2 - 3 | 187 | 91 | |
| JP055004.1 | - | 2 - 3 | 187 | 91 | ||
Set Membership: |
|