Search
Search

Gars| Gene Symbol: Gars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644462.1 | XP_007642652.1 | 13 - 14 | 1547 | 68 | |
| GenBank mRNA | JI895973.1 | - | 12 - 13 | 1453 | 68 | |
| JP039959.1 | - | 12 - 13 | 1453 | 68 | ||
Set Membership: |
|