Search
Search

Sep15| Gene Symbol: Sep15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256866.1 | NP_001243795.1 | 1 - 2 | 273 | 74 | |
| GenBank mRNA | JI893016.1 | - | 1 - 2 | 273 | 74 | |
| JP051733.1 | - | 1 - 2 | 273 | 74 | ||
Set Membership: |
|