Search
Search

Aimp1| Gene Symbol: Aimp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244267.1 | NP_001231196.1 | 444 | 82 | ||
| XM_007607757.1 | XP_007605947.1 | 124 | 82 | |||
| GenBank mRNA | AF021800.1 | - | 444 | 82 | ||
| JI863491.1 | - | 116 | 82 | |||
Set Membership: |
|