Search
Search

Znf330| Gene Symbol: Znf330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244298.1 | NP_001231227.1 | 5 - 6 | 567 | 82 | |
| GenBank mRNA | JI881202.1 | - | 2 - 3 | 312 | 82 | |
| JP065055.1 | - | 2 - 3 | 312 | 82 | ||
| Y12836.1 | - | 5 - 6 | 567 | 82 | ||