Search
Search

Sdhc| Gene Symbol: Sdhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246826.1 | NP_001233755.1 | 5 - 6 | 417 | 72 | |
| GenBank mRNA | JI877771.1 | - | 5 - 6 | 434 | 72 | |
| JP037986.1 | - | 5 - 6 | 434 | 72 | ||
| U31241.1 | - | 5 - 6 | 417 | 72 | ||
Set Membership: |
|