Search
Search

Orc5| Gene Symbol: Orc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271426.1 | NP_001258355.1 | 8 - 9 | 903 | 75 | |
| XM_007642868.1 | XP_007641058.1 | 8 - 9 | 926 | 75 | ||
| GenBank mRNA | AB183828.1 | - | 8 - 9 | 947 | 75 | |
| DQ016320.1 | - | 8 - 9 | 823 | 75 | ||
| JI889702.1 | - | 6 - 7 | 564 | 75 | ||
| JP041682.1 | - | 8 - 9 | 903 | 75 | ||
Set Membership: |
|