Search
Search

Abcb1b| Gene Symbol: Abcb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243989.1 | NP_001230918.1 | 22 - 23 | 2927 | 72 | |
| XM_007646774.1 | XP_007644964.1 | 23 - 24 | 3091 | 72 | ||
| XM_007646775.1 | XP_007644965.1 | 21 - 22 | 2866 | 72 | ||
| GenBank mRNA | M17896.1 | - | 9 - 10 | 1056 | 72 | |
| M60041.1 | - | 22 - 23 | 2927 | 72 | ||
Set Membership: |
|