Search
Search

G6pd| Gene Symbol: G6pd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246727.1 | NP_001233656.1 | 3 - 4 | 290 | 86 | |
| GenBank mRNA | AF044676.1 | - | 3 - 4 | 290 | 86 | |
| JF935143.1 | - | 2 - 3 | 155 | 86 | ||
| JI880982.1 | - | 3 - 4 | 269 | 86 | ||
| JP058833.1 | - | 3 - 4 | 269 | 86 | ||
Set Membership: |
|