Search
Search

Gnas| Gene Symbol: Gnas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244053.1 | NP_001230982.1 | 9 - 10 | 1062 | 64 | |
| GenBank mRNA | AF487552.1 | - | 9 - 10 | 1062 | 64 | |
| DQ202704.1 | - | 9 - 10 | 966 | 64 | ||
| JI883390.1 | - | 9 - 10 | 944 | 64 | ||
| JP041770.1 | - | 9 - 10 | 944 | 64 | ||
Set Membership: |
|