Search
Search

Smu1| Gene Symbol: Smu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244852.1 | NP_001231781.1 | 11 - 12 | 1438 | 95 | |
| XM_007651990.1 | XP_007650180.1 | 12 - 13 | 1854 | 95 | ||
| GenBank mRNA | AB111945.1 | - | 11 - 12 | 1438 | 95 | |
| JI870928.1 | - | 11 - 12 | 1495 | 95 | ||
| JP037369.1 | - | 8 - 9 | 981 | 95 | ||
Set Membership: |
|