Search
Search

Smu1| Gene Symbol: Smu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244852.1 | NP_001231781.1 | 8 - 9 | 1001 | 92 | |
| XM_007651990.1 | XP_007650180.1 | 9 - 10 | 1417 | 92 | ||
| GenBank mRNA | AB111945.1 | - | 8 - 9 | 1001 | 92 | |
| JI870928.1 | - | 8 - 9 | 1058 | 92 | ||
| JP037369.1 | - | 5 - 6 | 544 | 92 | ||