Search
Search

Cope| Gene Symbol: Cope | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246677.1 | NP_001233606.1 | 4 - 5 | 476 | 96 | |
| GenBank mRNA | JI891514.1 | - | 4 - 5 | 483 | 96 | |
| JP045922.1 | - | 4 - 5 | 483 | 96 | ||
| Z32554.1 | - | 4 - 5 | 476 | 96 | ||
Set Membership: |
|