Search
Search

Impdh2| Gene Symbol: Impdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246822.1 | NP_001233751.1 | 12 - 13 | 1467 | 89 | |
| XM_007652642.1 | XP_007650832.1 | 13 - 14 | 1581 | 89 | ||
| GenBank mRNA | J04209.1 | - | 12 - 13 | 1467 | 89 | |
| JI878814.1 | - | 3 - 4 | 436 | 89 | ||
| JP055329.1 | - | 3 - 4 | 436 | 89 | ||
Set Membership: |
|