Search
Search

LOC100774713| Gene Symbol: LOC100774713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512701.2 | XP_003512749.2 | 15 - 16 | 1398 | 68 | |
| XM_007653120.1 | XP_007651310.1 | 15 - 16 | 1426 | 68 | ||
| GenBank mRNA | JI882393.1 | - | 15 - 16 | 1294 | 68 | |
Set Membership: |
|