Search
Search

Ankrd55| Gene Symbol: Ankrd55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644035.1 | XP_007642225.1 | 2 - 3 | 241 | 61 | |
| XM_007644036.1 | XP_007642226.1 | 2 - 3 | 241 | 61 | ||
Set Membership: |
|