Search
Search

Pacsin2| Gene Symbol: Pacsin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648651.1 | XP_007646841.1 | 5 - 6 | 751 | 56 | |
| GenBank mRNA | JI888817.1 | - | 2 - 3 | 393 | 56 | |
| JP038067.1 | - | 4 - 5 | 705 | 56 | ||
Set Membership: |
|