Search
Search

Tp53bp1| Gene Symbol: Tp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007621707.1 | XP_007619897.1 | 9 - 10 | 1200 | 85 | |
| XM_007621713.1 | XP_007619903.1 | 9 - 10 | 1197 | 85 | ||
| XM_007621720.1 | XP_007619910.1 | 9 - 10 | 1200 | 85 | ||
| XM_007621728.1 | XP_007619918.1 | 9 - 10 | 1200 | 85 | ||
| GenBank mRNA | JI880814.1 | - | 6 - 7 | 769 | 85 | |
| JP044749.1 | - | 6 - 7 | 769 | 85 | ||
Set Membership: |
|