Search
Search

Evc2| Gene Symbol: Evc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651797.1 | XP_007649987.1 | 14 - 15 | 2481 | 63 | |
| XM_007651800.1 | XP_007649990.1 | 13 - 14 | 2436 | 63 | ||
Set Membership: |
|