Search
Search

Nipal3| Gene Symbol: Nipal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504926.2 | XP_003504974.1 | 3 - 4 | 595 | 53 | |
| XM_007645420.1 | XP_007643610.1 | 3 - 4 | 596 | 53 | ||
| XM_007645421.1 | XP_007643611.1 | 3 - 4 | 595 | 53 | ||
Set Membership: |
|