Search
Search

Actr6| Gene Symbol: Actr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507330.2 | XP_003507378.1 | 2 - 3 | 255 | 76 | |
| XM_007647893.1 | XP_007646083.1 | 2 - 3 | 256 | 76 | ||
| GenBank mRNA | JI886718.1 | - | 2 - 3 | 272 | 76 | |
| JP051281.1 | - | 2 - 3 | 272 | 76 | ||
Set Membership: |
|