Search
Search

Skiv2l2| Gene Symbol: Skiv2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503435.2 | XP_003503483.1 | 20 - 21 | 2542 | 67 | |
| GenBank mRNA | JI893126.1 | - | 20 - 21 | 2321 | 67 | |
| JP045169.1 | - | 20 - 21 | 2321 | 67 | ||
Set Membership: |
|