Search
Search

LOC100770743| Gene Symbol: LOC100770743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507962.2 | XP_003508010.1 | 10 - 11 | 2042 | 65 | |
| XM_007648575.1 | XP_007646765.1 | 10 - 11 | 2061 | 65 | ||
| XM_007648576.1 | XP_007646766.1 | 10 - 11 | 2055 | 65 | ||
| XM_007648577.1 | XP_007646767.1 | 10 - 11 | 2061 | 65 | ||
| XM_007648578.1 | XP_007646768.1 | 10 - 11 | 2324 | 65 | ||
| XM_007648579.1 | XP_007646769.1 | 9 - 10 | 1954 | 65 | ||
| XM_007648580.1 | XP_007646770.1 | 11 - 12 | 1950 | 65 | ||
| GenBank mRNA | JI888570.1 | - | 4 - 5 | 484 | 65 | |
Set Membership: |
|