Search
Search

Chrnd| Gene Symbol: Chrnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513313.1 | XP_003513361.1 | 4 - 5 | 357 | 63 | |
| XM_007653617.1 | XP_007651807.1 | 4 - 5 | 348 | 63 | ||
| XM_007653618.1 | XP_007651808.1 | 3 - 4 | 303 | 63 | ||
Set Membership: |
|