Search
Search

Cask| Gene Symbol: Cask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495368.2 | XP_003495416.1 | 4 - 5 | 367 | 69 | |
| XM_007640299.1 | XP_007638489.1 | 4 - 5 | 367 | 69 | ||
| XM_007640303.1 | XP_007638493.1 | 4 - 5 | 367 | 69 | ||
Set Membership: |
|