Search
Search

Nrd1| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496495.2 | XP_003496543.1 | 12 - 13 | 1699 | 113 | |
| XM_003496496.2 | XP_003496544.1 | 14 - 15 | 1905 | 113 | ||
| GenBank mRNA | JI877071.1 | - | 12 - 13 | 1691 | 113 | |
| JP058756.1 | - | 12 - 13 | 1691 | 113 | ||
Set Membership: |
|