Search
Search

Nrd1| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496495.2 | XP_003496543.1 | 18 - 19 | 2244 | 98 | |
| XM_003496496.2 | XP_003496544.1 | 20 - 21 | 2450 | 98 | ||
| GenBank mRNA | JI877071.1 | - | 18 - 19 | 2236 | 98 | |
| JP058756.1 | - | 18 - 19 | 2236 | 98 | ||
Set Membership: |
|