Search
Search

Sympk| Gene Symbol: Sympk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645981.1 | XP_007644171.1 | 701 | 55 | ||
| GenBank mRNA | JI883734.1 | - | 6 - 7 | 777 | 55 | |
| JP057020.1 | - | 6 - 7 | 777 | 55 | ||
Set Membership: |
|